accessibility menu, dialog, popup

UserName

CAS RN 394-33-2

OHFNOO

CAS RN 394-33-2

IUPAC Name: 4-fluoro-2-nitrophenol
Synonyms: phenol,2-nitro,4-fluoro
Molecular Weight (g/mol): 157.1
Molecular Formula: C6H4FNO3
InChi Key: ZHRLVDHMIJDWSS-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)