accessibility menu, dialog, popup

UserName

CAS RN 39332-13-3

NNHNNSOHOH

CAS RN 39332-13-3

IUPAC Name: 3-(7H-purin-6-ylsulfanyl)propane-1,2-diol
Synonyms: 3-(7H-purin-6-ylsulfanyl)propane-1,2-diol
Molecular Weight (g/mol): 226.25
Molecular Formula: C8H10N4O2S
InChi Key: OPKFIKAYNXEJQA-UHFFFAOYNA-N
SMILES: OCC(O)CSC1=C2NC=NC2=NC=N1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results