CAS RN 3913-67-5
CAS RN 3913-67-5
IUPAC Name:
(2S)-2-(methylamino)propanoic acid
Synonyms:
N-methylalanine
Molecular Weight (g/mol):
103.12
Molecular Formula:
C4H9NO2
InChi Key:
GDFAOVXKHJXLEI-VKHMYHEASA-N
SMILES:
CN[C@@H](C)C(O)=O
1
–
1
of
1
results
1
N-Methyl-L-alanine, 98%
CAS: 3913-67-5 Molecular Formula: C4H9NO2 Molecular Weight (g/mol): 103.121 MDL Number: MFCD00037241 InChI Key: GDFAOVXKHJXLEI-VKHMYHEASA-N Synonym: n-methyl-l-alanine,n-methylalanine,2s-2-methylamino propanoic acid,h-n-me-ala-oh,l-alanine, n-methyl,alanine, n-methyl,s-n-methylalanine,s-2-methylamino propanoic acid,n-,a-methyl-l-alanine,alanine, n-methyl-, l PubChem CID: 5288725 ChEBI: CHEBI:17519 IUPAC Name: (2S)-2-(methylamino)propanoic acid SMILES: CC(C(=O)O)NC