accessibility menu, dialog, popup

UserName

CAS RN 38585-74-9

HOSN

CAS RN 38585-74-9

IUPAC Name: (1,3-thiazol-5-yl)methanol
Synonyms: 1,3-thiazol-5-ylmethanol
Molecular Weight (g/mol): 115.15
Molecular Formula: C4H5NOS
InChi Key: WKBQQWDVVHGWDB-UHFFFAOYSA-N
SMILES: OCC1=CN=CS1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results