CAS RN 38465-60-0
CAS RN 38465-60-0
IUPAC Name:
copper(2+) bis(tetrafluoroboranuide)
Synonyms:
copper(II) tetrafluoroborate
Molecular Weight (g/mol):
237.15
Molecular Formula:
B2CuF8
InChi Key:
HMUNZEYTSRPVBE-UHFFFAOYSA-N
SMILES:
[Cu++].F[B-](F)(F)F.F[B-](F)(F)F