accessibility menu, dialog, popup

UserName

CAS RN 381247-99-0

CH3OONHOBr

CAS RN 381247-99-0

IUPAC Name: methyl 5-bromo-6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms: methyl 5-bromo-6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: KKMVACLRBBJYIJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CNC(=O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results