CAS RN 38048-32-7
CAS RN 38048-32-7
IUPAC Name:
2-(hydroxymethyl)-5-(6-{[(4-nitrophenyl)methyl]sulfanyl}-9H-purin-9-yl)oxolane-3,4-diol
Synonyms:
nitrobenzylthioinosine
Molecular Weight (g/mol):
419.41
Molecular Formula:
C17H17N5O6S
InChi Key:
DYCJFJRCWPVDHY-UHFFFAOYNA-N
SMILES:
OCC1OC(C(O)C1O)N1C=NC2=C(SCC3=CC=C(C=C3)[N+]([O-])=O)N=CN=C12
1
–
2
of
2
results
1
S-(4-Nitrobenzyl)-6-thioinosine, TRC
CAS: 38048-32-7 Chemical Name or Material: S-(4-Nitrobenzyl)-6-thioinosine Formula Weight: 419.09 InChI Formula: InChI=1S/C17H17N5O6S/c23-5-11-13(24)14(25)17(28-11)21-8-20-12-15(21)18-7-19-16(12)29-6-9-1-3-10(4-2-9)22(26)27/h1-4,7-8,11,13-14,17,23-25H,5-6H2/t11-,13-,14-,17-/m1/s1 IUPAC Name: (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[(4-nitrophenyl)methylsulfanyl]purin-9-yl]oxolane-3,4-diol Molecular Formula: C17 H17 N5 O6 S Molecular Weight (g/mol): 419.41 Recommended Storage: 4°C SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(SCc4ccc(cc4)[N+](=O)[O-])ncnc23