accessibility menu, dialog, popup

UserName

CAS RN 380315-80-0

H3COHNNHSNHOCH3H3CCH3

CAS RN 380315-80-0

IUPAC Name: N-[4-({[(4-tert-butylphenyl)formamido]methanethioyl}amino)phenyl]acetamide
Synonyms: N-[4-({[(4-tert-butylphenyl)formamido]methanethioyl}amino)phenyl]acetamide
Molecular Weight (g/mol): 369.48
Molecular Formula: C20H23N3O2S
InChi Key: WOWJIWFCOPZFGV-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(NC(=S)NC(=O)C2=CC=C(C=C2)C(C)(C)C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results