CAS RN 3800-06-4
CAS RN 3800-06-4
IUPAC Name:
2-(4-fluorobenzoyl)aniline
Synonyms:
2-(4-fluorobenzoyl)aniline
Molecular Weight (g/mol):
215.23
Molecular Formula:
C13H10FNO
InChi Key:
FFFXIQFESQNINT-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1C(=O)C1=CC=C(F)C=C1
1
–
2
of
2
results
1
2-Amino-4'-fluorobenzophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-4'-fluorobenzophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.