CAS RN 37945-06-5
CAS RN 37945-06-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-Chloro-2-(3-chloromethyl-4H-1,2,4-triazol-4-yl)-benzophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Chloro-2-(3-chloromethyl-4H-1,2,4-triazol-4-yl)-benzophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.