CAS RN 379231-04-6
CAS RN 379231-04-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Saracatinib, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Saracatinib, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Promotions Available
Tocris Bioscience™ Saracatinib
Tocris Bioscience™ Saracatinib
Potent and selective Src tyrosine kinase family inhibitor; anticancer; antiviral; orally bioavailable