accessibility menu, dialog, popup

UserName

CAS RN 3788-56-5

HOOO

CAS RN 3788-56-5

IUPAC Name: 9-hydroxynonanoate
Synonyms: 9-hydroxynonanoate
Molecular Weight (g/mol): 173.23
Molecular Formula: C9H17O3
InChi Key: AFZMICRBFKZNIH-UHFFFAOYSA-M
SMILES: OCCCCCCCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)