CAS RN 37812-51-4
CAS RN 37812-51-4
IUPAC Name:
4-[(4-cyanophenyl)methyl]morpholin-4-ium
Synonyms:
4-[(4-cyanophenyl)methyl]morpholin-4-ium
Molecular Weight (g/mol):
203.26
Molecular Formula:
C12H15N2O
InChi Key:
BCZLYNFDOJXWGN-UHFFFAOYSA-O
SMILES:
N#CC1=CC=C(C[NH+]2CCOCC2)C=C1