accessibility menu, dialog, popup

UserName

CAS RN 37693-18-8

ClOClO

CAS RN 37693-18-8

IUPAC Name: 4-chlorobutyl carbonochloridate
Synonyms: 4-chlorobutyl carbonochloridate
Molecular Weight (g/mol): 171.02
Molecular Formula: C5H8Cl2O2
InChi Key: OTDWRXBRYNVCDN-UHFFFAOYSA-N
SMILES: ClCCCCOC(Cl)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results