CAS RN 376608-74-1
CAS RN 376608-74-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-[[(3aR,4S,6R,6aS)-6-[[5-Amino-6-chloro-2-(propylthio)-4-pyrimidinyl]amino]tetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]oxy]ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[[(3aR,4S,6R,6aS)-6-[[5-Amino-6-chloro-2-(propylthio)-4-pyrimidinyl]amino]tetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]oxy]ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-[[(3aR,4S,6R,6aS)-6-[[5-Amino-6-chloro-2-(propylthio)-4-pyrimidinyl]amino]tetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]oxy]ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.