accessibility menu, dialog, popup

UserName

CAS RN 3766-55-0

H2NNHSNHCH2

CAS RN 3766-55-0

IUPAC Name: 1-amino-3-(prop-2-en-1-yl)thiourea
Synonyms: 1-amino-3-(prop-2-en-1-yl)thiourea
Molecular Weight (g/mol): 131.2
Molecular Formula: C4H9N3S
InChi Key: CZLPCLANGIXFIE-UHFFFAOYSA-N
SMILES: NNC(=S)NCC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results