CAS RN 376584-30-4
CAS RN 376584-30-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1(2H)-isoquinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.