CAS RN 3754-05-0
CAS RN 3754-05-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
16alpha,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
16Alpha,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
16Alpha,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.