CAS RN 37526-88-8
CAS RN 37526-88-8
IUPAC Name:
benzyl (2E)-2-methylbut-2-enoate
Synonyms:
benzyl tiglate
Molecular Weight (g/mol):
190.24
Molecular Formula:
C12H14O2
InChi Key:
QRGSTISKDZCDHV-XCVCLJGOSA-N
SMILES:
C\C=C(/C)C(=O)OCC1=CC=CC=C1
1
–
2
of
2
results
1
Tiglic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tiglic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.