CAS RN 3748-13-8
CAS RN 3748-13-8
IUPAC Name:
1,3-bis(prop-1-en-2-yl)benzene
Synonyms:
1,3-bis(prop-1-en-2-yl)benzene
Molecular Weight (g/mol):
158.24
Molecular Formula:
C12H14
InChi Key:
IBVPVTPPYGGAEL-UHFFFAOYSA-N
SMILES:
CC(=C)C1=CC(=CC=C1)C(C)=C