accessibility menu, dialog, popup

UserName

CAS RN 374633-31-5

CH3NNOOBr

CAS RN 374633-31-5

IUPAC Name: 2-bromo-6-methyl-3-nitropyridine
Synonyms: 2-bromo-6-methyl-3-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: IHHLJCDGOSZFTB-UHFFFAOYSA-N
SMILES: CC1=NC(Br)=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results