CAS RN 3740-92-9
CAS RN 3740-92-9
IUPAC Name:
4,6-dichloro-2-phenylpyrimidine
Synonyms:
fenclorim
Molecular Weight (g/mol):
225.07
Molecular Formula:
C10H6Cl2N2
InChi Key:
NRFQZTCQAYEXEE-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=NC(=N1)C1=CC=CC=C1
1
–
2
of
2
results
1
Fenclorim, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fenclorim, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.