CAS RN 3737-09-5
CAS RN 3737-09-5
IUPAC Name:
4-[bis(propan-2-yl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide
Synonyms:
disopyramide disopiramida disopyramidum
Molecular Weight (g/mol):
339.48
Molecular Formula:
C21H29N3O
InChi Key:
UVTNFZQICZKOEM-UHFFFAOYNA-N
SMILES:
CC(C)N(CCC(C(N)=O)(C1=CC=CC=C1)C1=CC=CC=N1)C(C)C
1
–
3
of
3
results
1
Disopyramide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Disopyramide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Disopyramide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.