CAS RN 37299-86-8
CAS RN 37299-86-8
IUPAC Name:
disodium 9-(2,4-dicarboxylatophenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Synonyms:
α-methylphenethylamine
Molecular Weight (g/mol):
566.99
Molecular Formula:
C29H29ClN2Na2O5
InChi Key:
NRZDMKVYRRMFRR-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=C(C=C3C([O-])=O)C([O-])=O)=C2C=C1)N(CC)CC
1
–
1
of
1
results
1
Thermo Scientific Chemicals Rhodamine WT, 20% solution in water
CAS: 37299-86-8 | C29H29ClN2Na2O5 | 566.99 g/mol