CAS RN 372963-49-0
CAS RN 372963-49-0
IUPAC Name:
(5-methylpyridin-2-yl)boronic acid
Synonyms:
5-methylpyridin-2-ylboronic acid
Molecular Weight (g/mol):
136.95
Molecular Formula:
C6H8BNO2
InChi Key:
VDRIXWWGBDNWCM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(N=C1)B(O)O