CAS RN 372941-54-3
CAS RN 372941-54-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.