accessibility menu, dialog, popup

UserName

CAS RN 372198-97-5

CH3NOFFFOOH

CAS RN 372198-97-5

IUPAC Name: 2-[methyl({3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl})amino]acetic acid
Synonyms: [methyl({3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl})amino]acetic acid
Molecular Weight (g/mol): 367.37
Molecular Formula: C19H20F3NO3
InChi Key: KZWQAWBTWNPFPW-UHFFFAOYNA-N
SMILES: CN(CCC(OC1=CC=C(C=C1)C(F)(F)F)C1=CC=CC=C1)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results