CAS RN 372-66-7
CAS RN 372-66-7
IUPAC Name:
hydrogen 6-amino-2-methylheptan-2-ol chloride
Synonyms:
heptaminol
Molecular Weight (g/mol):
181.7
Molecular Formula:
C8H20ClNO
InChi Key:
JZNBMCOSOXIZJB-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(N)CCCC(C)(C)O
1
–
3
of
3
results
1
Heptaminol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Heptaminol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Heptaminol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.