CAS RN 371775-73-4
CAS RN 371775-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
tert-Butyl-7-[4-(4-fluorophenyl)-6-isopropyl-2-mesylaminopyrimidin-5-yl]-(3R,5S)-isopropylidine-(E)-6-heptenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl-7-[4-(4-fluorophenyl)-6-isopropyl-2-mesylaminopyrimidin-5-yl]-(3R,5S)-isopropylidine-(E)-6-heptenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.