accessibility menu, dialog, popup

UserName

CAS RN 37063-35-7

N(R)N(R)O(R)(R)NNNH2HOOPOOONa

CAS RN 37063-35-7

IUPAC Name: sodium 4-(6-amino-9H-purin-9-yl)-2-hydroxy-6-(hydroxymethyl)-tetrahydro-2H-2λ⁵-furo[3,4-d][1,3,2]dioxaphosphol-2-one hydride
Synonyms: sodium 4-(6-aminopurin-9-yl)-2-hydroxy-6-(hydroxymethyl)-tetrahydro-2λ⁵-furo[3,4-d][1,3,2]dioxaphosphol-2-one hydride
Molecular Weight (g/mol): 353.21
Molecular Formula: C10H13N5NaO6P
InChi Key: IKFVEGHTFODLHX-UHFFFAOYNA-N
SMILES: [H-].[Na+].NC1=C2N=CN(C3OC(CO)C4OP(O)(=O)OC34)C2=NC=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)