CAS RN 36923-21-4
CAS RN 36923-21-4
IUPAC Name:
diphenylmethyl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms:
diphenylmethyl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol):
366.44
Molecular Formula:
C20H18N2O3S
InChi Key:
NYYBPASOTOAXQW-UHFFFAOYNA-N
SMILES:
NC1C2SCC=C(N2C1=O)C(=O)OC(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Diphenylmethyl 7Beta-Amino-3-cephem-4-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.