CAS RN 3691-12-1
CAS RN 3691-12-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Guaiene (>85%), TRC
CAS: 3691-12-1 Molecular Formula: C15H24 Molecular Weight (g/mol): 204.35 Synonym: Guaia-1(5),11-diene,(1S,4S,7R)-1,2,3,4,5,6,7,8-Octahydro-1,4-dimethyl-7-(1-methylethenyl)azulene,1,2,3,4,5,6,7,8-Octahydro-1,4-dimethyl-7-(1-methylethenyl)-[1S-(1alpha,4alpha,7alpha)] azulene IUPAC Name: (1S,4S,7R)-1,4-dimethyl-7-prop-1-en-2-yl-1,2,3,4,5,6,7,8-octahydroazulene SMILES: C[C@H]1CC[C@H](CC2=C1CC[C@@H]2C)C(=C)C