accessibility menu, dialog, popup

UserName

CAS RN 3685-22-1

OHOOH

CAS RN 3685-22-1

IUPAC Name: 4-hydroxycyclohexane-1-carboxylic acid
Synonyms: 4-hydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: HCFRWBBJISAZNK-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)