accessibility menu, dialog, popup

UserName

H3CCH2OOFF(S)FFFF

CAS RN 36405-47-7

IUPAC Name: (3S)-2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
Synonyms: (3S)-2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 250.14
Molecular Formula: C8H8F6O2
InChi Key: DFVPUWGVOPDJTC-LURJTMIESA-N
SMILES: CC(=C)C(=O)OCC(F)(F)[C@H](F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results