accessibility menu, dialog, popup

UserName

CAS RN 36138-76-8

CH3BrOH

CAS RN 36138-76-8

IUPAC Name: 5-bromo-2-methylphenol
Synonyms: 5-bromo-2-methylphenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: OONJCAWRVJDVBB-UHFFFAOYSA-N
SMILES: CC1=C(O)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results