accessibility menu, dialog, popup

UserName

CAS RN 3544-24-9

H2NOH2N

CAS RN 3544-24-9

IUPAC Name: 3-aminobenzamide
Synonyms: 3-aminobenzamide 3-H2NC6H4CONH2
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: GSCPDZHWVNUUFI-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)