accessibility menu, dialog, popup

UserName

CAS RN 352530-29-1

CHNHCl

CAS RN 352530-29-1

IUPAC Name: 4-ethynylpyridine hydrochloride
Synonyms: 4-ethynylpyridine hydrochloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C7H6ClN
InChi Key: KFBZWZGIZHLUBX-UHFFFAOYSA-N
SMILES: Cl.C#CC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)