accessibility menu, dialog, popup

UserName

CAS RN 3495-35-0

RbOO

CAS RN 3495-35-0

IUPAC Name: rubidium(1+) formate
Synonyms: rubidium(1+) formate
Molecular Weight (g/mol): 130.49
Molecular Formula: CHO2Rb
InChi Key: ZIMBPNXOLRMVGV-UHFFFAOYSA-M
SMILES: [Rb+].[O-]C=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results