accessibility menu, dialog, popup

UserName

CH3ONH2FFF

CAS RN 349-55-3

IUPAC Name: 3-methoxy-5-(trifluoromethyl)aniline
Synonyms: 3-methoxy-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: VTFGJEYZCUWSAM-UHFFFAOYSA-N
SMILES: COC1=CC(N)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results