accessibility menu, dialog, popup

UserName

CAS RN 34841-06-0

H3COOBr

CAS RN 34841-06-0

IUPAC Name: 3-bromo-4-methoxybenzaldehyde
Synonyms: 3-bromo-4-methoxybenzaldehyde
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: QMPNFQLVIGPNEI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)