CAS RN 3482-57-3
CAS RN 3482-57-3
IUPAC Name:
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
463.39
Molecular Formula:
C18H25NO13
InChi Key:
IAYJZWFYUSNIPN-KFRZSCGFSA-N
SMILES:
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OC3=CC=C(C=C3)[N+]([O-])=O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
1
–
3
of
3
results
1
p-Nitrophenyl Beta-D-Cellobioside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
p-Nitrophenyl Beta-D-Cellobioside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
p-Nitrophenyl Beta-D-Cellobioside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.