accessibility menu, dialog, popup

UserName

CAS RN 34688-71-6

CH3CH3NH3C

CAS RN 34688-71-6

IUPAC Name: 2-(2,4,6-trimethylphenyl)acetonitrile
Synonyms: 2-(2,4,6-trimethylphenyl)acetonitrile
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: SDKQOGSGNPGPRN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CC#N)C(C)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results