accessibility menu, dialog, popup

UserName

CAS RN 346719-56-0

IUPAC Name: 1-(2-fluorobenzoyl)-4-methylpiperidine
Synonyms: 1-(2-fluorobenzoyl)-4-methylpiperidine
Molecular Weight (g/mol): 221.28
Molecular Formula: C13H16FNO
InChi Key: ZUOUPMRPDVWTSP-UHFFFAOYSA-N
SMILES: CC1CCN(CC1)C(=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results