accessibility menu, dialog, popup

UserName

H3CONFFF

CAS RN 34636-92-5

IUPAC Name: 2-methoxy-5-(trifluoromethyl)benzonitrile
Synonyms: 2-methoxy-5-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: KAJZODLWVHKBSI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C#N)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results