CAS RN 34562-31-7
IUPAC Name:
3,5-diethyl-1-phenyl-2-propyl-1,2-dihydropyridine
Synonyms:
3,5-diethyl-1-phenyl-2-propyl-2H-pyridine
Molecular Weight (g/mol):
255.41
Molecular Formula:
C18H25N
InChi Key:
IAAASXBHFUJLHW-UHFFFAOYNA-N
SMILES:
CCCC1N(C=C(CC)C=C1CC)C1=CC=CC=C1
1
–
1
of
1
results
1
3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, tech. 75%, Thermo Scientific™
CAS: 34562-31-7 Molecular Formula: C18H25N Molecular Weight (g/mol): 255.41 MDL Number: MFCD00071789 InChI Key: IAAASXBHFUJLHW-UHFFFAOYNA-N Synonym: 3,5-diethyl-1-phenyl-2-propyl-1,2-dihydropyridine,3,5-diethyl-1,2-dihydro-1-phenyl-2-propylpyridine,pdhp,pyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl,1-phenyl-3,5-diethyl-2-propyl-1,2-dihydropyridine,dsstox_cid_24923,dsstox_rid_80589,dsstox_gsid_44923,3,5-diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine,3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl pyridine PubChem CID: 118703 IUPAC Name: 3,5-diethyl-1-phenyl-2-propyl-2H-pyridine SMILES: CCCC1N(C=C(CC)C=C1CC)C1=CC=CC=C1