accessibility menu, dialog, popup

UserName

HClH3COHNOOHHNCH3CH3CH3O

CAS RN 34381-68-5

IUPAC Name: hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride
Synonyms: hydrogen acebutolol chloride
Molecular Weight (g/mol): 372.89
Molecular Formula: C18H29ClN2O4
InChi Key: KTUFKADDDORSSI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O
Certifications/Compliance 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)