CAS RN 34283-69-7
CAS RN 34283-69-7
IUPAC Name:
caesium(1+) vanadium trioxidandiide
Synonyms:
caesium(1+) vanadium trioxidandiide
Molecular Weight (g/mol):
231.85
Molecular Formula:
CsO3V
InChi Key:
WWENEIGBGOIMMI-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[V].[Cs+]
1
–
1
of
1
results
1
Cesium orthovanadate, 99.9% (metals basis)
CAS: 34283-69-7 MDL Number: MFCD00210609 Synonym: cesium orthovanadate,tricaesium 1+ vanadate 3-