accessibility menu, dialog, popup

UserName

CAS RN 3387-41-5

H3CCH3CH2

CAS RN 3387-41-5

IUPAC Name: 4-methylidene-1-(propan-2-yl)bicyclo[3.1.0]hexane
Synonyms: sabinene sabinen 4(10)-thujene (1R)-sabinene (1r)-sabinene sabinene, (1R)- sabinene, (1r)- sabinene, (1R)-isomer sabinene, (1r)-isomer (1R)-isomer sabinene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: NDVASEGYNIMXJL-UHFFFAOYNA-N
SMILES: CC(C)C12CC1C(=C)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results