CAS RN 3383-21-9
CAS RN 3383-21-9
IUPAC Name:
3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione
Synonyms:
3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol):
220.31
Molecular Formula:
C14H20O2
InChi Key:
NOUZOVBGCDDMSX-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C
1
–
2
of
2
results
1
3,5-Di-tert-butyl-o-benzoquinone, 98+%
CAS: 3383-21-9 Molecular Formula: C14H20O2 Molecular Weight (g/mol): 220.312 MDL Number: MFCD00001647 InChI Key: NOUZOVBGCDDMSX-UHFFFAOYSA-N Synonym: 3,5-di-tert-butyl-o-benzoquinone,3,5-di-tert-butyl-1,2-benzoquinone,3,5-di tert-butyl benzo-1,2-quinone,3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl,3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione,o-benzoquinone, 3,5-di-tert-butyl,3,5-di-tert-butylbenzo-1,2-quinone,3,5-di-t-butyl-1,2-benzoquinone,3,5-ditert-butylbenzo-1,2-quinone,3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione PubChem CID: 76915 IUPAC Name: 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione SMILES: CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C