accessibility menu, dialog, popup

UserName

CAS RN 336100-47-1

H3COOOHNH2HCl

CAS RN 336100-47-1

IUPAC Name: 2-amino-3-methoxypropanoic acid hydrochloride
Synonyms: DL-serine, O-methyl- hydrochloride
Molecular Weight (g/mol): 155.58
Molecular Formula: C4H10ClNO3
InChi Key: NLDVBPFPOCWZCE-UHFFFAOYNA-N
SMILES: Cl.COCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)